4-Fluorzimtsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(p-Fluorphenyl)propensäure, 4-Fluorcinnamsäure, 3-(4-Fluorphenyl)propensäure
001180-25G
001180-100G
001180-250G
FLUO001180-25GEA
45.9
EUR
FLUO001180-25G
FLUO001180-100G
FLUO001180-250G
4-Fluorzimtsäure
4-Fluorzimtsäure
Formel:
C₉H₇FO₂ Schmelzpunkt: 209 °C Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00004395 CAS-Nummer: 459-32-5 EINECS: 207-288-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...