1-Fluor-2-iodbenzol
Lieferant: FLUOROCHEM
Synonyme:
2-Fluoroiodobenzene
001182-100G
001182-250G
FLUO001182-100GEA
143
EUR
FLUO001182-100G
FLUO001182-250G
1-Fluor-2-iodbenzol
1-Fluor-2-iodbenzol
Formel:
C₆H₄FI Molecular Weight: 222 g/mol Siedepunkt: 188 Schmelzpunkt: –41.5 °C Dichte: 1.903 g/cm³ (20 °C) |
MDL:
MFCD00001031 CAS-Nummer: 348-52-7 EINECS: 206-477-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...