(2-Fluorphenyl)acetonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Fluorbenzylcyanid

001187.25G 001187.100G
FLUO001187.25GEA 44.1 EUR
FLUO001187.25G FLUO001187.100G
(2-Fluorphenyl)acetonitril
(2-Fluorphenyl)acetonitril
Formel: C₈H₆FN
Molecular Weight: 135.14 g/mol
Siedepunkt: 114
Dichte: 1.14045 g/cm³ (15 °C)
MDL: MFCD00001897
CAS-Nummer: 326-62-5
EINECS: 206-313-0
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR