5'-Fluor-2'-hydroxyacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(5-Fluoro-2-hydroxyphenyl)ethanone
001235-1G
001235-5G
001235-25G
FLUO001235-1GEA
0
EUR
FLUO001235-1G
FLUO001235-5G
FLUO001235-25G
5'-Fluor-2'-hydroxyacetophenon
5'-Fluor-2'-hydroxyacetophenon
Formel:
C₈H₇FO₂ Molecular Weight: 154.14 g/mol Siedepunkt: 182 Schmelzpunkt: 56 |
MDL:
MFCD00011668 CAS-Nummer: 394-32-1 EINECS: 206-893-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...