3-Acetyl-2,5-dichlorthiophen

Lieferant: FLUOROCHEM

Synonyme: 1-(2,5-Dichlor-3-thienyl)-1-ethanon

001362.5G 001362-25G
FLUO001362.5GEA 33 EUR
FLUO001362.5G FLUO001362-25G
3-Acetyl-2,5-dichlorthiophen
3-Acetyl-2,5-dichlorthiophen
Formel: C₆H₄Cl₂OS
Molecular Weight: 195,07 g/mol
Siedepunkt: 120…122 °C (6 mmHg)
Schmelzpunkt: 37…40 °C
MDL: MFCD00014522
CAS-Nummer: 36157-40-1
EINECS: 252-893-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR