2-Chlorbenzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (2-Chlorophenyl)methanol

001375-100G
FLUO001375-100GEA 79.1 EUR
FLUO001375-100G
2-Chlorbenzylalkohol
2-Chlorbenzylalkohol
Formel: C₇H₇ClO
Molecular Weight: 142,58 g/mol
Siedepunkt: 227 °C (1013 hPa)
Schmelzpunkt: 68…71 °C
MDL: MFCD00004604
CAS-Nummer: 17849-38-6
EINECS: 241-801-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR