4'-Trifluormethoxyacetophenon
Lieferant: FLUOROCHEM
001407.1G
001407-5G
001407-100G
001407-25G
FLUO001407.1GEA
0
EUR
FLUO001407.1G
FLUO001407-5G
FLUO001407-100G
FLUO001407-25G
4'-Trifluormethoxyacetophenon
4'-Trifluormethoxyacetophenon
Formel:
C₉H₇F₃O₂ Siedepunkt: 110 °C (30 mmHg) Dichte: 1,27 g/cm³ (20 °C) |
MDL:
MFCD00042404 CAS-Nummer: 85013-98-5 EINECS: 285-066-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...