1,1,1-Trifluoriodethan
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2,2,2-Trifluoriodethan, 2-Iod-1,1,1-trifluorethan, 1-Iod-2,2,2-trifluorethan, 2,2,2-Trifluorethyliodid
001536-25G
FLUO001536-25GEA
119
EUR
FLUO001536-25G
1,1,1-Trifluoriodethan
1,1,1-Trifluoriodethan
Formel:
C₂H₂F₃I Molecular Weight: 209,94 g/mol Siedepunkt: 54,7 °C (1013 hPa) Dichte: 2,1499 g/cm³ (20 °C) |
MDL:
MFCD00001082 CAS-Nummer: 353-83-3 EINECS: 206-541-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...