3,3,3-Trifluor-1-propanol
Lieferant: FLUOROCHEM
Synonyme:
3,3,3-Trifluoropropan-1-ol
001557-1G
001557-5G
FLUO001557-1GEA
42.9
EUR
FLUO001557-1G
FLUO001557-5G
3,3,3-Trifluor-1-propanol
3,3,3-Trifluor-1-propanol
Formel:
C₃H₅F₃O Molecular Weight: 114.07 g/mol Siedepunkt: 96…98 °C (1013 hPa) Dichte: 1.3739 g/cm³ (25 °C) |
MDL:
MFCD00190649 CAS-Nummer: 2240-88-2 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...