α,α,α,α',α',α'-Hexafluor-m-xylol
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
α,α,α,β,β,β-Hexafluor-m-xylol
001600-1KG
001600-250G
FLUO001600-1KGEA
203
EUR
FLUO001600-1KG
FLUO001600-250G
α,α,α,α',α',α'-Hexafluor-m-xylol
α,α,α,α',α',α'-Hexafluor-m-xylol
Formel:
C₈H₄F₆ Molecular Weight: 214.11 g/mol Siedepunkt: 116,3 Schmelzpunkt: –35 °C Dichte: 1.383 g/cm³ (22 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000392 CAS-Nummer: 402-31-3 EINECS: 206-939-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...