1-Fluor-2-phenoxybenzol

Lieferant: FLUOROCHEM

001609-1G
FLUO001609-1GEA 321 EUR
FLUO001609-1G
1-Fluor-2-phenoxybenzol
1-Fluoro-2-phenoxybenzene
Formel: C₁₂H₉FO
MDL: MFCD00236610
CAS-Nummer: 124330-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR