1,2-Difluor-4-nitrobenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3,4-Difluornitrobenzol
001664-25G
001664-100G
001664-500G
FLUO001664-25GEA
31.1
EUR
FLUO001664-25G
FLUO001664-100G
FLUO001664-500G
1,2-Difluor-4-nitrobenzol
1,2-Difluor-4-nitrobenzol
Formel:
C₆H₃F₂NO₂ Molecular Weight: 159.09 g/mol Siedepunkt: 80 Dichte: 1.43 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007198 CAS-Nummer: 369-34-6 EINECS: 206-718-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...