3-Chlorthiophen

Lieferant: FLUOROCHEM

001747-25G 001747-100G
FLUO001747-25GEA 72.9 EUR
FLUO001747-25G FLUO001747-100G
3-Chlorthiophen
3-Chlorthiophen
Formel: C₄H₃ClS
Molecular Weight: 118.59 g/mol
Siedepunkt: 133…135 °C (1013 hPa)
Dichte: 1.276 g/cm³ (20 °C)
MDL: MFCD00043887
CAS-Nummer: 17249-80-8
UN: 1992
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR