4-Methoxythiophenol
Lieferant: FLUOROCHEM
Synonyme:
p-Methoxythiophenol, 4-Mercaptoanisol, 4-Methoxybenzol-1-thiol, 1-Methoxy-4-mercaptobenzol, 4-Methoxybenzolthiol, 4-Methoxyphenylmercaptan
001774-25G
001774-10G
FLUO001774-25GEA
76.2
EUR
FLUO001774-25G
FLUO001774-10G
4-Methoxythiophenol
4-Methoxythiophenol
Formel:
C₇H₈OS Molecular Weight: 140.21 g/mol Siedepunkt: 228 Schmelzpunkt: 42 Dichte: 1.1397 g/cm³ (20 °C) |
MDL:
MFCD00004849 CAS-Nummer: 696-63-9 EINECS: 211-799-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...