2,3,5-Trifluorphenol

Lieferant: FLUOROCHEM

001834-1G
FLUO001834-1GEA 395 EUR
FLUO001834-1G
2,3,5-Trifluorphenol
2,3,5-Trifluorphenol
Formel: C₆H₃F₃O
Molecular Weight: 148,08 g/mol
MDL: MFCD00075416
CAS-Nummer: 2268-15-7
UN: 1325
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR