2,3-Difluoranilin
Lieferant: FLUOROCHEM
001855-5G
001855-25G
001855-100G
FLUO001855-5GEA
40.2
EUR
FLUO001855-5G
FLUO001855-25G
FLUO001855-100G
2,3-Difluoranilin
2,3-Difluoranilin
Formel:
C₆H₅F₂N Molecular Weight: 129.11 g/mol Siedepunkt: 69 °C (12 mmHg) Dichte: 1.274 g/cm³ (20 °C) |
MDL:
MFCD00010298 CAS-Nummer: 4519-40-8 EINECS: 224-847-2 UN: 2941 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...