2,3-Difluorbenzonitril
Lieferant: FLUOROCHEM
001859-5G
001859-25G
FLUO001859-5GEA
36.9
EUR
FLUO001859-5G
FLUO001859-25G
2,3-Difluorbenzonitril
2,3-Difluorbenzonitril
Formel:
C₇H₃F₂N Siedepunkt: 189 °C (1013 hPa) Dichte: 1,25 g/cm³ (20 °C) |
MDL:
MFCD00009976 CAS-Nummer: 21524-39-0 EINECS: 244-418-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...