2,3-Difluorphenol
Lieferant: FLUOROCHEM
001865-5G
001865-25G
FLUO001865-5GEA
30.6
EUR
FLUO001865-5G
FLUO001865-25G
2,3-Difluorphenol
2,3-Difluorphenol
Formel:
C₆H₄F₂O Siedepunkt: 54 °C (25 mmHg) Schmelzpunkt: 30…32 °C |
MDL:
MFCD00010262 CAS-Nummer: 6418-38-8 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...