2,4-Difluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
2,4-Difluorbenzoesäureamid
001916-25G
FLUO001916-25GEA
104
EUR
FLUO001916-25G
2,4-Difluorbenzamid
2,4-Difluorbenzamid
Formel:
C₇H₅F₂NO Molecular Weight: 157,12 g/mol |
MDL:
MFCD00015547 CAS-Nummer: 85118-02-1 EINECS: 285-654-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...