2',6'-Difluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2,6-Difluorphenyl)ethan-1-on

001954-5G 001954-25G 001954-100G
FLUO001954-5GEA 0 EUR
FLUO001954-5G FLUO001954-25G FLUO001954-100G
2',6'-Difluoracetophenon
2',6'-Difluoracetophenon
Formel: C₈H₆F₂O
Molecular Weight: 156.13 g/mol
Siedepunkt: 76…79 °C (15 mmHg)
Dichte: 1.19 g/cm³ (20 °C)
MDL: MFCD00000328
CAS-Nummer: 13670-99-0
EINECS: 237-151-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR