2-Fluor-6-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
Synonyme:
2-Amino-α,α,α,3-tetrafluortoluol, α,α,α,6-Tetrafluor-o-toluidin
001982-5G
001982-25G
FLUO001982-5GEA
183
EUR
FLUO001982-5G
FLUO001982-25G
2-Fluor-6-(trifluormethyl)anilin
2-Fluor-6-(trifluormethyl)anilin
Formel:
C₇H₅F₄N Molecular Weight: 179.12 g/mol Siedepunkt: 155 Dichte: 1.388 g/cm³ (20 °C) |
MDL:
MFCD00061242 CAS-Nummer: 144851-61-6 UN: 1992 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...