4-Brom-2-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
Synonyme:
2-Amino-5-brom-α,α,α-trifluortoluol, 4-Brom-α,α,α-trifluor-o-toluidin
001983-25G
001983-100G
FLUO001983-25GEA
39.9
EUR
FLUO001983-25G
FLUO001983-100G
4-Brom-2-(trifluormethyl)anilin
4-Brom-2-(trifluormethyl)anilin
Formel:
C₇H₅BrF₃N Siedepunkt: 60,7…62 °C (1 mmHg) Dichte: 1,712 g/cm³ (25 °C) |
MDL:
MFCD00064393 CAS-Nummer: 445-02-3 EINECS: 207-150-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...