3,4-Difluorbenzonitril
Lieferant: FLUOROCHEM
002081-25G
002081-100G
FLUO002081-25GEA
35.3
EUR
FLUO002081-25G
FLUO002081-100G
3,4-Difluorbenzonitril
3,4-Difluorbenzonitril
Formel:
C₇H₃F₂N Siedepunkt: 189 °C (1013 hPa) Schmelzpunkt: 51 °C |
MDL:
MFCD00011666 CAS-Nummer: 64248-62-0 EINECS: 264-751-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...