3',5'-Bis(trifluormethyl)acetophenon
Lieferant: FLUOROCHEM
002095-10G
002095-250G
002095-50G
FLUO002095-10GEA
45.9
EUR
FLUO002095-10G
FLUO002095-250G
FLUO002095-50G
3',5'-Bis(trifluormethyl)acetophenon
3',5'-Bis(trifluormethyl)acetophenon
Formel:
C₁₀H₆F₆O Siedepunkt: 95 °C (15 mmHg) Dichte: 1,42 g/cm³ (20 °C) |
MDL:
MFCD00009910 CAS-Nummer: 30071-93-3 EINECS: 250-023-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...