3-(Trifluormethoxy)benzylalkohol
Lieferant: FLUOROCHEM
002125-1G
002125-5G
002125-10G
002125-25G
FLUO002125-1GEA
0
EUR
FLUO002125-1G
FLUO002125-5G
FLUO002125-10G
FLUO002125-25G
3-(Trifluormethoxy)benzylalkohol
3-(Trifluormethoxy)benzylalkohol
Formel:
C₈H₇F₃O₂ |
MDL:
MFCD00061270 CAS-Nummer: 50823-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...