3-(Trifluormethoxy)benzylalkohol

Lieferant: FLUOROCHEM

002125-1G 002125-5G 002125-10G 002125-25G
FLUO002125-1GEA 0 EUR
FLUO002125-1G FLUO002125-5G FLUO002125-10G FLUO002125-25G
3-(Trifluormethoxy)benzylalkohol
3-(Trifluormethoxy)benzylalkohol
Formel: C₈H₇F₃O₂
MDL: MFCD00061270
CAS-Nummer: 50823-90-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR