3-(Trifluormethyl)benzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
(3-(Trifluoromethyl)phenyl)methanol, 3-(Hydroxymethyl)benzotrifluoride
002135-10G
002135-100G
002135-25G
FLUO002135-10GEA
0
EUR
FLUO002135-10G
FLUO002135-100G
FLUO002135-25G
3-(Trifluormethyl)benzylalkohol
3-(Trifluormethyl)benzylalkohol
Formel:
C₈H₇F₃O Molecular Weight: 176.14 g/mol Siedepunkt: 68 Dichte: 1.2949 g/cm³ (21 °C) |
MDL:
MFCD00004645 CAS-Nummer: 349-75-7 EINECS: 206-489-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...