3-(Trifluormethyl)benzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (3-(Trifluoromethyl)phenyl)methanol, 3-(Hydroxymethyl)benzotrifluoride

002135-10G 002135-100G 002135-25G
FLUO002135-10GEA 0 EUR
FLUO002135-10G FLUO002135-100G FLUO002135-25G
3-(Trifluormethyl)benzylalkohol
3-(Trifluormethyl)benzylalkohol
Formel: C₈H₇F₃O
Molecular Weight: 176.14 g/mol
Siedepunkt: 68
Dichte: 1.2949 g/cm³ (21 °C)
MDL: MFCD00004645
CAS-Nummer: 349-75-7
EINECS: 206-489-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR