2-Chlor-1-fluor-4-nitrobenzol
Lieferant: FLUOROCHEM
002152-100G
002152-500G
002152-50G
FLUO002152-100GEA
51.9
EUR
FLUO002152-100G
FLUO002152-500G
FLUO002152-50G
2-Chlor-1-fluor-4-nitrobenzol
2-Chlor-1-fluor-4-nitrobenzol
Formel:
C₆H₃ClFNO₂ Siedepunkt: 227…228 °C (1013 hPa) Schmelzpunkt: 41,5 °C |
MDL:
MFCD00007206 CAS-Nummer: 350-30-1 EINECS: 206-499-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...