3-Fluor-5-(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

002160-1G 002160-5G 002160-25G
FLUO002160-1GEA 31.1 EUR
FLUO002160-1G FLUO002160-5G FLUO002160-25G
3-Fluor-5-(trifluormethyl)benzonitril
3-Fluor-5-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
Molecular Weight: 189.11 g/mol
Siedepunkt: 212
Schmelzpunkt: 28
MDL: MFCD00061282
CAS-Nummer: 149793-69-1
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR