4-(Trifluormethoxy)benzylalkohol
Lieferant: FLUOROCHEM
002175-1G
002175-5G
002175-25G
FLUO002175-1GEA
31.1
EUR
FLUO002175-1G
FLUO002175-5G
FLUO002175-25G
4-(Trifluormethoxy)benzylalkohol
4-(Trifluormethoxy)benzylalkohol
Formel:
C₈H₇F₃O₂ Siedepunkt: 53…54 °C (0,5 mmHg) Dichte: 1,32 g/cm³ (20 °C) |
MDL:
MFCD00036029 CAS-Nummer: 1736-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...