4-Chlor-3-fluorbenzaldehyd

Lieferant: FLUOROCHEM
Achtung

002198-5G 002198-25G
FLUO002198-5GEA 0 EUR
FLUO002198-5G FLUO002198-25G
4-Chlor-3-fluorbenzaldehyd
4-Chloro-3-fluorobenzaldehyde
Formel: C₇H₄ClFO
MDL: MFCD00143288
CAS-Nummer: 5527-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR