α,α,α,4-Tetrafluor-m-toluylsäure

Lieferant: FLUOROCHEM

CFF632.2.5 CFF632.5 CFF632.10 002222-5G 002222-25G 002222-1.5KG 002222-100G
CHMPCFF632.2.5EA 268 EUR
CHMPCFF632.2.5 CHMPCFF632.5 CHMPCFF632.10 FLUO002222-5G FLUO002222-25G FLUO002222-1.5KG FLUO002222-100G
α,α,α,4-Tetrafluor-m-toluylsäure
α,α,α,4-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208.11 g/mol
MDL: MFCD00061294
CAS-Nummer: 67515-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR