5-Brom-α,α,α-trifluor-2-nitrotoluol
Lieferant: FLUOROCHEM
Synonyme:
1-Brom-4-nitro-3-(trifluormethyl)benzol, 5-Brom-2-nitrobenzotrifluorid
002247.10G
002247.50G
002247-25G
002247-100G
FLUO002247.10GEA
24.2
EUR
FLUO002247.10G
FLUO002247.50G
FLUO002247-25G
FLUO002247-100G
5-Brom-α,α,α-trifluor-2-nitrotoluol
5-Brom-α,α,α-trifluor-2-nitrotoluol
Formel:
C₇H₃BrF₃NO₂ Molecular Weight: 270.01 g/mol Siedepunkt: 95…100 °C (5 mmHg) Schmelzpunkt: 40…45 °C Dichte: 1.799 g/cm³ (25 °C) |
MDL:
MFCD00014706 CAS-Nummer: 344-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...