1H,1H,9H,9H-Perfluor-1,9-nonandiol
Lieferant: FLUOROCHEM
Synonyme:
2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluor-1,9-nonandiol
002306-5G
002306-25G
FLUO002306-5GEA
72.9
EUR
FLUO002306-5G
FLUO002306-25G
1H,1H,9H,9H-Perfluor-1,9-nonandiol
1H,1H,9H,9H-Perfluor-1,9-nonandiol
Formel:
C₉H₆F₁₄O₂ Molecular Weight: 412.12 g/mol |
MDL:
MFCD00153275 CAS-Nummer: 203303-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...