1H,1H,10H,10H-Hexadecafluor-1,10-decandiol

Lieferant: FLUOROCHEM

002307-5G 002307-25G
FLUO002307-5GEA 84.9 EUR
FLUO002307-5G FLUO002307-25G
1H,1H,10H,10H-Hexadecafluor-1,10-decandiol
1H,1H,10H,10H-Hexadecafluor-1,10-decandiol
Formel: C₁₀H₆F₁₆O₂
MDL: MFCD00192200
CAS-Nummer: 754-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR