1H,1H,12H,12H-Eicosafluor-1,12-dodecandiol

Lieferant: FLUOROCHEM

002308-5G 002308-25G 002308-1G
FLUO002308-5GEA 115 EUR
FLUO002308-5G FLUO002308-25G FLUO002308-1G
1H,1H,12H,12H-Eicosafluor-1,12-dodecandiol
1H,1H,12H,12H-Eicosafluor-1,12-dodecandiol
Formel: C₁₂H₆F₂₀O₂
MDL: MFCD00236625
CAS-Nummer: 183162-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR