2-Fluor-5-nitrotoluol
Lieferant: FLUOROCHEM
002351-25G
002351-100G
002351-250G
FLUO002351-25GEA
30.8
EUR
FLUO002351-25G
FLUO002351-100G
FLUO002351-250G
2-Fluor-5-nitrotoluol
2-Fluor-5-nitrotoluol
Formel:
C₇H₆FNO₂ Siedepunkt: 230 °C (1013 hPa) Schmelzpunkt: 40 °C |
MDL:
MFCD00007284 CAS-Nummer: 455-88-9 EINECS: 207-251-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...