Bis(4-fluorphenyl)sulfon

Lieferant: FLUOROCHEM

Synonyme: 4,4'-Difluordiphenylsulfon, 4-Fluorphenylsulfon

002477-25G 002477-100G
FLUO002477-25GEA 51.8 EUR
FLUO002477-25G FLUO002477-100G
Bis(4-fluorphenyl)sulfon
Bis(4-fluorphenyl)sulfon
Formel: C₁₂H₈F₂O₂S
Molecular Weight: 254.26 g/mol
MDL: MFCD00000350
CAS-Nummer: 383-29-9
EINECS: 206-847-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR