6-Amino-3-brom-2-methylpyridin
Lieferant: FLUOROCHEM
|
Achtung
|
002495-1G
002495-5G
002495-25G
002495.25
FLUO002495-1GEA
31.1
EUR
FLUO002495-1G
FLUO002495-5G
FLUO002495-25G
FLUO002495.25
6-Amino-3-brom-2-methylpyridin
6-Amino-3-brom-2-methylpyridin
Formel:
C₆H₇BrN₂ |
MDL:
MFCD00068230 CAS-Nummer: 42753-71-9 EINECS: 255-927-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...