6-Amino-3-brom-2-methylpyridin

Lieferant: FLUOROCHEM
Achtung

002495-1G 002495-5G 002495-25G 002495.25
FLUO002495-1GEA 31.1 EUR
FLUO002495-1G FLUO002495-5G FLUO002495-25G FLUO002495.25
6-Amino-3-brom-2-methylpyridin
6-Amino-3-brom-2-methylpyridin
Formel: C₆H₇BrN₂
MDL: MFCD00068230
CAS-Nummer: 42753-71-9
EINECS: 255-927-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR