3-(3-Aminophenyl)propionsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(3-Aminophenyl)propanoic acid
002499-5G
FLUO002499-5GEA
162
EUR
FLUO002499-5G
3-(3-Aminophenyl)propionsäure
3-(3-Aminophenyl)propionsäure
Formel:
C₉H₁₁NO₂ |
MDL:
MFCD00153889 CAS-Nummer: 1664-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...