3-(4-Aminophenyl)propionsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(4-Aminophenyl)propansäure

002500-5G 002500-25G
FLUO002500-5GEA 189 EUR
FLUO002500-5G FLUO002500-25G
3-(4-Aminophenyl)propionsäure
3-(4-Aminophenyl)propionsäure
Formel: C₉H₁₁NO₂
MDL: MFCD00017118
CAS-Nummer: 2393-17-1
EINECS: 219-245-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR