2',5'-Dimethylacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2,5-Dimethylphenyl)ethan-1-on
002546-25G
FLUO002546-25GEA
0
EUR
FLUO002546-25G
2',5'-Dimethylacetophenon
2',5'-Dimethylacetophenon
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 231 °C (1013 hPa) Schmelzpunkt: –18 °C Dichte: 1,0154 g/cm³ (0 °C) |
MDL:
MFCD00014988 CAS-Nummer: 2142-73-6 EINECS: 218-399-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...