2-Iod-5-methylthiophen

Lieferant: FLUOROCHEM

002582-5G
FLUO002582-5GEA 124 EUR
FLUO002582-5G
2-Iod-5-methylthiophen
2-Iod-5-methylthiophen
Formel: C₅H₅IS
Lagertemperatur: Kühlschrank
MDL: MFCD00022492
CAS-Nummer: 16494-36-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR