4-Methoxy-β-nitrostyrol

Lieferant: FLUOROCHEM

Synonyme: trans-4-Methoxy-β-nitrostyrol

002591-5G
FLUO002591-5GEA 130 EUR
FLUO002591-5G
4-Methoxy-β-nitrostyrol
4-Methoxy-β-nitrostyrol
Formel: C₉H₉NO₃
MDL: MFCD00024826
CAS-Nummer: 3179-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR