Phloretin

Lieferant: FLUOROCHEM

002612-1G
FLUO002612-1GEA 39.9 EUR
FLUO002612-1G
Phloretin
Phloretin
Formel: C₁₅H₁₄O₅
Molecular Weight: 274,28 g/mol
MDL: MFCD00002288
CAS-Nummer: 60-82-2
EINECS: 200-488-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR