3-(Trifluormethyl)phenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: α,α,α-Trifluoro-m-tolyl isothiocyanate

002619-25G 002619-10G
FLUO002619-25GEA 359 EUR
FLUO002619-25G FLUO002619-10G
3-(Trifluormethyl)phenylisothiocyanat
3-(Trifluormethyl)phenylisothiocyanat
Formel: C₈H₄F₃NS
Siedepunkt: 206…208 °C (1013 hPa)
Dichte: 1,33 g/cm³ (20 °C)
MDL: MFCD00004806
CAS-Nummer: 1840-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR