3-(Trifluormethyl)thiophenol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-(Trifluormethyl)phenylmercaptan, 3-(Trifluormethyl)benzolthiol, 3-Mercaptobenzotrifluorid

002621-5G 002621-25G
FLUO002621-5GEA 84.9 EUR
FLUO002621-5G FLUO002621-25G
3-(Trifluormethyl)thiophenol
3-(Trifluormethyl)thiophenol
Formel: C₇H₅F₃S
Molecular Weight: 178.18 g/mol
Siedepunkt: 161…163 °C (1013 hPa)
MDL: MFCD00041142
CAS-Nummer: 937-00-8
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR