4'-(Trifluormethyl)acetanilid

Lieferant: FLUOROCHEM

Synonyme: N-(4-(Trifluormethyl)phenyl)acetamid, Ethansäure(4-(trifluormethyl)phenyl)amid, 4-Acetamidobenzotrifluorid

002700-5G 002700-25G
FLUO002700-5GEA 45.9 EUR
FLUO002700-5G FLUO002700-25G
4'-(Trifluormethyl)acetanilid
4'-(Trifluormethyl)acetanilid
Formel: C₉H₈F₃NO
Molecular Weight: 203.16 g/mol
Schmelzpunkt: 208…211 °C
MDL: MFCD00013562
CAS-Nummer: 349-97-3
EINECS: 206-495-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR